Support rods, polished
12 mm, 500 mm
Catalog # RSGA241.205
Supplier: RSG
Jump to:
Product Details & Documents
Polished rods made from stainless steel 18/10. Available with or without thread.
Specifications
- Ø:12 mm
- Length:500 mm
- Seq:09
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



