Spatulas, micro
Powder spatula, 150 mm, 5 mm
RSGA087.375
: RSG
Stainless steel, 18/10. Micro spatulas with black plastic handle.
- Description:Powder spatula
- Length:150 mm
- Blade width:5 mm
Stainless steel, 18/10. Micro spatulas with black plastic handle.