Orders LinkContactUsLinkComponent
Forceps, straight
Forceps, straight
Fine tips, 200 mm
Catalog # 232-0076
Supplier:   RSG
Forceps, straight
Catalog # 232-0076
Supplier:  RSG
Forceps, straight
Forceps, straight
Fine tips, 200 mm
Price
97,40 €
Log in for availability
1 * 5 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
  • Serrated tips
  • With guide pin
Specifications
  • Type:
    Fine tips
  • Length:
    200 mm