Orders LinkContactUsLinkComponent
Forceps with guide pin
Forceps with guide pin
Curved, with guide pin, 105 mm
Catalog # 232-0077
Supplier:   RSG
Forceps with guide pin
Catalog # 232-0077
Supplier:  RSG
Forceps with guide pin
Forceps with guide pin
Curved, with guide pin, 105 mm
Price
54,90 €
Log in for availability
1 * 5 ST
Product Details & Documents
Stainless steel, anti-magnetic. Particularly suitable for staining applications.
  • With curved, extra fine tips
  • Corrosion resistant
Specifications
  • Type:
    Curved, with guide pin
  • Length:
    105 mm