Order Entry
Belgium
Orders LinkContactUsLinkComponent
Double spatulas, micro, stiff

Double spatulas, micro, stiff

100 mm, 5 mm
Catalog # RSGA835.100
Supplier:   RSG
Double spatulas, micro, stiff
Catalog # RSGA835.100
Supplier:  RSG
Double spatulas, micro, stiff
Double spatulas, micro, stiff

100 mm, 5 mm

Price
22,50 €
Log in for availability
1 * 5 ST
Product Details & Documents

Rigid micro double spatulas made of stainless steel.

Specifications
  • Length:
    100 mm
  • Blade width:
    5 mm