Order Entry
Belgium
Orders LinkContactUsLinkComponent
Double spatulas, micro, stiff

Double spatulas, micro, stiff

185 mm, 6 mm
Catalog # RSGA836.185
Supplier:   RSG
Double spatulas, micro, stiff
Catalog # RSGA836.185
Supplier:  RSG
Double spatulas, micro, stiff
Double spatulas, micro, stiff

185 mm, 6 mm

Price
24,70 €
Log in for availability
1 * 5 ST
Product Details & Documents

Rigid micro double spatulas made of stainless steel.

Specifications
  • Length:
    185 mm
  • Blade width:
    6 mm