Orders LinkContactUsLinkComponent
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Straight, blunt, 160 mm
Catalog # RSGA011.160
Supplier:   RSG
Forceps, premium quality
Catalog # RSGA011.160
Supplier:  RSG
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Straight, blunt, 160 mm
Price
48,90 €
Log in for availability
1 * 5 ST
Product Details & Documents
Stainless steel 1,4301.
Specifications
  • Type:
    Straight, blunt
  • Description:
    Forceps, premium quality
  • Length:
    160 mm