Orders LinkContactUsLinkComponent
Forceps, premium quality
Forceps, premium quality, Curved, blunt, 200 mm
  RSGA012.200
 :   RSG
Forceps, premium quality
  RSGA012.200
 :  RSG
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Curved, blunt, 200 mm
89,90 €
1 * 5 ST
Stainless steel 1,4301.
  • Type:
    Curved, blunt
  • Description:
    Forceps, premium quality
  • Length:
    200 mm