Orders LinkContactUsLinkComponent
Dishes
Dishes
Set, 90/115/120 mm, 175/217/245 mm
Catalog # 216-1249
Supplier:   RSG
Dishes
Catalog # 216-1249
Supplier:  RSG
Dishes
Dishes
Set, 90/115/120 mm, 175/217/245 mm
Price
73,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, ¹⁸/₁₀.
  • With folded edge; fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
Round multipurpose dishes for a wide range of uses.
Informations de commande: Each dish set (Cat. No. 216-1249) comprises one of each 1, 2 and 3 litre dishes.
Specifications
  • Ext.Ø:
    175/217/245 mm
  • Capacity:
    Set
  • Height:
    90/115/120 mm
  • Seq:
    10