Order Entry
Belgium
Orders LinkContactUsLinkComponent
Double spatulas, narrow

Double spatulas, narrow

Stiff, 350 mm, 16 mm
Catalog # RSGA830.350
Supplier:   RSG
Double spatulas, narrow
Catalog # RSGA830.350
Supplier:  RSG
Double spatulas, narrow
Double spatulas, narrow

Stiff, 350 mm, 16 mm

Price
55,20 €
Log in for availability
1 * 5 ST
Product Details & Documents

Double-ended, narrow spatulas made from stainless steel. Available flexible or non flexibe.

Specifications
  • Description:
    Stiff
  • Length:
    350 mm
  • Blade width:
    16 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.