Dishes
5 l, 135 mm, 315 mm
216-2023
: RSG
Stainless steel, ¹⁸/₁₀.
- With folded edge; fast, easy cleaning
- Polished electrolytically, anti-magnetic
- Good chemical resistance, very good thermal resistance
Round multipurpose dishes for a wide range of uses.
: Each dish set (Cat. No. 216-1249) comprises one of each 1, 2 and 3 litre dishes.
- Ext.Ø:315 mm
- Capacity:5 l
- Height:135 mm
- Seq:06



