Orders LinkContactUsLinkComponent
Inoculating loops, reusable
Loops, length 75 mm, Ø 1,5 mm
  631-0084
 :   RSG
Inoculating loops, reusable
  631-0084
 :  RSG
Inoculating loops, reusable
Inoculating loops, reusable
Loops, length 75 mm, Ø 1,5 mm
15,40 €
1 * 10 ST
These NiCr 8020 loops are available in a range of sizes and lengths with internal diameter of 1,5, 2,5 or 4 mm. They are heat resistant and can be flame sterilised. Diameter of wire: 0,6 mm. Loop holder available separately.
  • Description:
    Loops, length 75 mm, Ø 1,5 mm
Accessories
  •  
    Description
    Length
  • 391-0374
    Needle holder
    240 mm
    1 * 1 ST - 15,00 €
  •  
    Description
    Length
  • 391-0373
    Needle holder
    170 mm
    1 * 1 ST - 13,00 €