Handheld refractometers
28 - 62 °Brix, 0,2 °Brix
N° de catalogue 635-0311
Fournisseur: EUROMEX
Jump to:
Product Details & Documents
Single dual or triple scales, analogue, handheld refractometers.
- Automated temperature compensation (ATC)
- Calibrated at 20 °C except for RF.5190
- Requires only a few drops of liquid
Specifications
- Range:28 - 62 °Brix
- Seq:04
- Precision:0,2 °Brix