Orders LinkContactUsLinkComponent
Test sets
Test sets
Test set for conductive compounds
N° de catalogue DRAE8103170
Fournisseur:   Dräger
Test sets
N° de catalogue DRAE8103170
Fournisseur:  Dräger
Test sets
Test sets
Test set for conductive compounds
Prix
135,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifications
  • Description:
    Test set for conductive compounds
  • Seq:
    04
  • Cat. No.:
    DRAE8103170
  • Model:
    Test set for conductive compounds
  • Measurement time:
    120 s