Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sample cuvettes

Flow through cuvette, micro, CH = 8,5 mm, 80 µl, 10 mm, Optical glass
  HACHLZP334
 :   Hach
Sample cuvettes
  HACHLZP334
 :  Hach
Sample cuvettes
Sample cuvettes

Flow through cuvette, micro, CH = 8,5 mm, 80 µl, 10 mm, Optical glass

 
 
   
1 * 1 Pce
 

Sample cuvettes for photometric measurements.

  • Available with or without lid
  • Available reusable (made from glass) or for single use (made from plastics)
 
  • Path length:
    10 mm
  • Volume:
    80 µl
  • Window material:
    Optical glass
  • Description:
    Flow through cuvette, micro, CH = 8,5 mm
  • Seq:
    12