Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer, single-use, SINGLET™

pH buffer, single-use, SINGLET™

pH buffer solution combination pack, single-use, 7,00/10,01, Foil pouch, 20/pack (10 each of 7,00 pH and 10,01 pH), 1 SET
Catalog # HACH2769820
Supplier:   Hach
pH buffer, single-use, SINGLET™
Catalog # HACH2769820
Supplier:  Hach
CAS Number:  
pH buffer, single-use, SINGLET™
pH buffer, single-use, SINGLET™

pH buffer solution combination pack, single-use, 7,00/10,01, Foil pouch, 20/pack (10 each of 7,00 pH and 10,01 pH), 1 SET

CAS Number:  
Price
50,70 €
Log in for availability
1 * 1 SET
Product Details & Documents

These buffers solutions packs are used for pH meter calibration. The sealed foil pouches can be transported easily and safely without any spillage and also prevents contamination or exposure to air of the solutions.

  • Ideal for on-site analyses
  • Easy to use
  • Accuracy pH: ±0,02 at 25 °C
  • Traceable to NIST

Documents

Specifications

Specifications

  • Pack type:
    Foil pouch
  • Pk:
    1 SET
  • Description:
    pH buffer solution combination pack, single-use
  • Pk Info:
    20/pack (10 each of 7,00 pH and 10,01 pH)
  • pH value:
    7,00/10,01

Specifications