Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, colour coded

pH buffer solutions, colour coded

Buffer solution, 7,00, ±0,02, Phosphate, Yellow, 25 °C, 500 mL
Catalog # HACH2283549.99
Supplier:   Hach
pH buffer solutions, colour coded
Catalog # HACH2283549.99
Supplier:  Hach
CAS Number:  
pH buffer solutions, colour coded
pH buffer solutions, colour coded

Buffer solution, 7,00, ±0,02, Phosphate, Yellow, 25 °C, 500 mL

CAS Number:  
Price
24,70 €
Log in for availability
1 * 500 mL
Product Details & Documents

For calibration of pH meters

  • Colour coded for easy identification
  • Accuracy (pH): ±0,02 at 25 °C

Certifications: Traceable to NIST.

Specifications

Specifications

  • pH buffer type:
    Phosphate
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Yellow
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,02
  • pH value:
    7,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.