Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer powder pillows

Buffer powder pillows

4,01, ±0,02, Phthalate, Red, 25 °C, 250
Catalog # HACH2226964
Supplier:   Hach
Buffer powder pillows
Catalog # HACH2226964
Supplier:  Hach
CAS Number:  
Buffer powder pillows
Buffer powder pillows

4,01, ±0,02, Phthalate, Red, 25 °C, 250

CAS Number:  
Price
286,00 €
Log in for availability
1 * 250 ST
Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

Delivery information: The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

Specifications

Specifications

  • pH buffer type:
    Phthalate
  • Pk:
    250
  • Reference temperature:
    25 °C
  • Colour:
    Red
  • pH accuracy:
    ±0,02
  • pH value:
    4,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.