Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH Buffer solutions

pH Buffer solutions

Buffer solution, 4,01, Colourless, 500 mL
  HACH1222349
 :   Hach
pH Buffer solutions
  HACH1222349
 :  Hach
 :  
pH Buffer solutions
pH Buffer solutions

Buffer solution, 4,01, Colourless, 500 mL

 :  
 
23,00 €
   
1 * 500 mL
 

For calibration of pH meters.

Accuracy: pH is ±0,02 at 25 °C

Certifications: Traceable to NIST.

Caution: Please note that the pH 4 and 10 buffers contain formaldehyde <0,1% and the pH 7 buffer contains formaldehyde 0,1 to 1%.

 

 

  • Pk:
    500 mL
  • Colour:
    Colourless
  • Description:
    Buffer solution
  • pH value:
    4,01

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.