Order Entry
Belgium
Orders LinkContactUsLinkComponent
TLC introductory kit, Micro Set A

TLC introductory kit, Micro Set A

Test dye mixture 1, solution of 4 lipophilic dyes in toluene, 8 ml
Catalog # MANA814001
Supplier:   MACHEREY-NAGEL
TLC introductory kit, Micro Set A
Catalog # MANA814001
Supplier:  MACHEREY-NAGEL
TLC introductory kit, Micro Set A
TLC introductory kit, Micro Set A

Test dye mixture 1, solution of 4 lipophilic dyes in toluene, 8 ml

Price
28,30 €
Log in for availability
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

An ideal introductory kit for the science education sector. The kit contains all the the chemicals, plates and accessories for students to perform separations of dyes including: anthaquinone dye mixtures, lipophilic dye mixtures and food dye mixtures. The TLC plates are polyester sheet based with either silica, cellulose or alumina as coating.

Delivery information: Kit includes 3 developing tanks, 50 glass capillaries (1 µl), 1 spotting guide, 1×10 ml measuring cylinder, 50 sheets 40×80 mm POLYGRAM SIL G/UV₂₅₄, 50 sheets 40×80 mm ALOX N/UV₂₅₄ and 50 sheets 40×80 mm of CEL 300, 3 dye mixtures (8 ml), 3 prepared solvent mixtures (100 ml), 2 felt pens and instruction manual.

Specifications
  • Description:
    Test dye mixture 1, solution of 4 lipophilic dyes in toluene, 8 ml
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.