Order Entry
Belgium
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Nickel

QUANTOFIX® Test Strips, Nickel

QUANTOFIX® multistick for aquarium owners, 0 • 6,3 • 12,5 • 18,8 • 25,0 • 31,3 - °e; 0 • 4 • 8 • 13 • 19 • 25 - °e; 6,4 • 6,8 • 7,2 • 7,6 • 8,0 • 8,4, Total hardness: color reaction with complex indicator, 40×40×120 mm (packaging), 100 Tests, 100 Tests
Catalog # MANA91326
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Nickel
Catalog # MANA91326
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Nickel
QUANTOFIX® Test Strips, Nickel

QUANTOFIX® multistick for aquarium owners, 0 • 6,3 • 12,5 • 18,8 • 25,0 • 31,3 - °e; 0 • 4 • 8 • 13 • 19 • 25 - °e; 6,4 • 6,8 • 7,2 • 7,6 • 8,0 • 8,4, Total hardness: color reaction with complex indicator, 40×40×120 mm (packaging), 100 Tests, 100 Tests

CAS Number:  
Price
49,10 €
Log in for availability
1 * 1 EA
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. QUANTOFIX® test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips are for the rapid and reliable determination of Nickel in solutions. The easy dip-and-read-procedure provides a reliable result within 30 seconds.

Specifications

Specifications

  • Chemical method:
    Total hardness: color reaction with complex indicator
  • Pk:
    100 Tests
  • Description:
    QUANTOFIX® multistick for aquarium owners
  • Dimensions:
    40×40×120 mm (packaging)
  • Tests per kit:
    100 Tests
  • Range:
    0 • 6,3 • 12,5 • 18,8 • 25,0 • 31,3 - °e; 0 • 4 • 8 • 13 • 19 • 25 - °e; 6,4 • 6,8 • 7,2 • 7,6 • 8,0 • 8,4
  • Cat. No.:
    MANA91326
  • Parameters:
    All reagents included
  • Contents:
    100 test strips in a tube, instruction leaflet
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    No
  • Shelf life (months):
    30 (from production)
  • Method:
    Carbonate hardness - Mixed indicator (bright green → blue);pH - mixed indicator (yellow → red);Total hardness - EDTA (green → red)
  • Weight:
    45,4 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.