Order Entry
Belgium
Orders LinkContactUsLinkComponent
Celite® 545, filter aid

Celite® 545, filter aid

5 kg
Catalog # MANA815560.5
Health
Supplier:   MACHEREY-NAGEL
Celite® 545, filter aid
Catalog # MANA815560.5
Supplier:  MACHEREY-NAGEL
CAS Number:  68855-54-9
Celite® 545, filter aid
Celite® 545, filter aid

5 kg

CAS Number:  68855-54-9
Price
149,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  • EINECS:272-489-0
Specifications

Specifications

  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.