VISOCOLOR® Colorimetric Test, Copper
VISOCOLOR® ECO Copper, refill pack, test no. 5-37; Measuring range on PF-12Plus. Range on other photometers can be different., Reagents for 100 tests in a box, 0 • 0,1 • 0,2 • 0,3 • 0,5 • 0,7 • 1,0 • 1.5 mg/L (Cu²⁺); 0,1 - 5,0 mg/L (Cu²⁺), Cuprizon, PF-12Plus;PF-3 Fish;Advance;Visual, 170×115×70 mm (Including packaging), 100, 1 KIT
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Reagents for the determination of copper in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: No
Specifications
Specifications
- Chemical method:Cuprizon
- Pk:1 KIT
- Description:VISOCOLOR® ECO Copper, refill pack, test no. 5-37; Measuring range on PF-12Plus. Range on other photometers can be different.
- Dimensions:170×115×70 mm (Including packaging)
- For unit:PF-12Plus;PF-3 Fish;Advance;Visual
- Tests per kit:100
- Range:0 • 0,1 • 0,2 • 0,3 • 0,5 • 0,7 • 1,0 • 1.5 mg/L (Cu²⁺); 0,1 - 5,0 mg/L (Cu²⁺)
- Contents:Reagents for 100 tests in a box
- Cat. No.:MANA931237
- Platform:Colorimetric test kits
- Shelf life (months):24
- Storage temperature:15...25 °C
- Weight:131,8 g (Including packaging)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



