Order Entry
Belgium
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

48×230 mm, 2,0 mm
Catalog # MANA645035
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Catalog # MANA645035
Supplier:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles

48×230 mm, 2,0 mm

Price
261,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.

  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifications
  • Int.Ø×H:
    48×230 mm
  • Seq:
    35
  • Thickness:
    2,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.