Order Entry
Belgium
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

28×120 mm, 1,5 mm
Catalog # MANA645014
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Catalog # MANA645014
Supplier:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles

28×120 mm, 1,5 mm

Price
117,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.

  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifications
  • Int.Ø×H:
    28×120 mm
  • Seq:
    15
  • Thickness:
    1,5 mm