Order Entry
Belgium
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

27× 80 mm, 1,5 mm
N° de catalogue MANA645009
Fournisseur:   MACHEREY-NAGEL
Extraction thimbles
N° de catalogue MANA645009
Fournisseur:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles

27× 80 mm, 1,5 mm

Prix
97,70 €
Connexion Pour la disponibilité
1 * 25 ST
Product Details & Documents

Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.

  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Spécifications
  • Int.Ø×H:
    27× 80 mm
  • Seq:
    10
  • Thickness:
    1,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.