Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, triethylammonium acetate, 1 M solution, pH 7,0

Buffer, triethylammonium acetate, 1 M solution, pH 7,0

Triethylammonium acetate, 1M buffer solution, pH 7,0, 1 L
Catalog # A3846.1000
Supplier:   PanReac AppliChem
Buffer, triethylammonium acetate, 1 M solution, pH 7,0
Catalog # A3846.1000
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, triethylammonium acetate, 1 M solution, pH 7,0
Buffer, triethylammonium acetate, 1 M solution, pH 7,0

Triethylammonium acetate, 1M buffer solution, pH 7,0, 1 L

CAS Number:  
Price
164,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Used for the purification of oligonucleotides.

CAS No.: 5204-74-0
Molecular Formula: C₆H₁₅N · CH₃COOH
Formula Weight: 161,24 g/mol
Density: 1,001 (at 20 °C)
Physical Form: Liquid
Non-volatile Matter: Max. 0,01%
Storage Temperature: Ambient

Documents

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Triethylammonium acetate, 1M buffer solution, pH 7,0
  • Cat. No.:
    A3846.1000
  • CAS No.:
    5204-74-0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.