Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Sucrose

D-(+)-Sucrose

Glass bottle for solids, 1 kg
Catalog # A2211.1000
Supplier:   PanReac AppliChem
D-(+)-Sucrose
Catalog # A2211.1000
Supplier:  PanReac AppliChem
CAS Number:  57-50-1
D-(+)-Sucrose
D-(+)-Sucrose

Glass bottle for solids, 1 kg

CAS Number:  57-50-1
Price
65,10 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS Number:57-50-1
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Melting Pt:169…170 °C
  • Density:1.29 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-334-9

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.