Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium deoxycholate

Sodium deoxycholate

Glass bottle for solids, 100 g
Catalog # A1531.0100
Irritant
Supplier:   PanReac AppliChem
Sodium deoxycholate
Catalog # A1531.0100
Supplier:  PanReac AppliChem
CAS Number:  302-95-4
Sodium deoxycholate

Glass bottle for solids, 100 g

CAS Number:  302-95-4
Price
144,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS Number:302-95-4
  • Formula:C₂₄H₃₉NaO₄
  • MDL Number:MFCD00064139
  • Molecular Weight:414.56 g/mol
  • Melting Pt:357…365 °C
  • Storage Temperature:Ambient
  • EINECS:206-132-7

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.