Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Galactose

D-(+)-Galactose

Glass bottle for solids, 1 kg
Catalog # A1131.1000
Supplier:   PanReac AppliChem
D-(+)-Galactose
Catalog # A1131.1000
Supplier:  PanReac AppliChem
CAS Number:  59-23-4
D-(+)-Galactose
D-(+)-Galactose

Glass bottle for solids, 1 kg

CAS Number:  59-23-4
Price
319,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:163…169 °C
  • Density:1.5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.