Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Ethidium bromide solution 0,07 %

Dropping bottle, 15 mL
Catalog # A2273.0015
Supplier:   PanReac AppliChem
Ethidium bromide solution 0,07 %
Catalog # A2273.0015
Supplier:  PanReac AppliChem
CAS Number:  1239-45-8
Ethidium bromide solution 0,07 %

Dropping bottle, 15 mL

CAS Number:  1239-45-8
Price
77,90 €
Log in for availability
1 * 15 mL
Product Details & Documents

Ethidium bromide solutions with a concentration of 0,7 mg/ml.

 : Supplied in a dropper bottle.

Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Dropping bottle
  • Pk:
    15 mL

Specifications