Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 100 mL
Catalog # A4892.0100
Supplier:   PanReac AppliChem
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Catalog # A4892.0100
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 100 mL

CAS Number:  
Price
35,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    EDTA buffer solution pH 8,0 (0,5 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.