Orders LinkContactUsLinkComponent
ATC probes, Eutech
ATC probes, Eutech
ATC probe, 1 m cable, pH 5+ / pH 6+ / ION 6+ meters
Catalog # EUTEECPH5TEM01P
Supplier:   EUTECH
ATC probes, Eutech
Catalog # EUTEECPH5TEM01P
Supplier:  EUTECH
ATC probes, Eutech
ATC probes, Eutech
ATC probe, 1 m cable, pH 5+ / pH 6+ / ION 6+ meters
Price
72,05 €
Log in for availability
1 * 1 ST
Product Details & Documents
ATC electrodes for use with Eutech pH 5+, pH 6+ and ION 6+ meters or for use with CyberScan pH 300 and 310 meters.
Specifications
  • For:
    pH 5+ / pH 6+ / ION 6+ meters
  • Description:
    ATC probe, 1 m cable