Orders LinkContactUsLinkComponent
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
10/30, 40 mm
Catalog # COWI010.110
Supplier:   COWIE
Stoppers, flat type, PTFE
Catalog # COWI010.110
Supplier:  COWIE
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
10/30, 40 mm
Price
10,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure
Specifications
  • NS:
    10/30
  • Height:
    40 mm