Orders LinkContactUsLinkComponent
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
19/38, 50 mm
Catalog # COWI010.119
Supplier:   COWIE
Stoppers, flat type, PTFE
Catalog # COWI010.119
Supplier:  COWIE
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
19/38, 50 mm
Price
17,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure
Specifications
  • NS:
    19/38
  • Height:
    50 mm