Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Removable needles

22, 0,72 mm, 0,41 mm, 5 or AS
  HAMI7731-03
Removable needles
  HAMI7731-03
Removable needles
Removable needles

22, 0,72 mm, 0,41 mm, 5 or AS

 
435,00 €
   
1 * 6 ST
 

The benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line. The result is a zero dead volume connection that simplifies priming.

Custom lengths and point styles are available. You can order your custom made needle according to your specification.

* Large hub

** Kel-F hub

 
  • Ext.Ø:
    0,72 mm
  • Gauge:
    22
  • Ø int.:
    0,41 mm
  • Type of tip:
    5 or AS
  • Seq:
    58