Order Entry
Belgium
Orders LinkContactUsLinkComponent
Counting chambers

Counting chambers

Counting chamber, dark lined, Fuchs-Rosenthal, 0,2 mm
Catalog # 630-1510
Supplier:   MARIENFELD
Counting chambers
Catalog # 630-1510
Supplier:  MARIENFELD
Counting chambers
Counting chambers

Counting chamber, dark lined, Fuchs-Rosenthal, 0,2 mm

Price
51,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

These counting chambers are suitable for counting leucocytes and erythrocytes.

  • Double net ruling
  • Two counting grids with one division
  • Different types of design available

 : IVD 98/79/EC.

 : Supplied with two cover glasses of 0,4 mm thickness.

Documents

Specifications
  • Depth of chamber:
    0,2 mm
  • Type:
    Fuchs-Rosenthal
  • Description:
    Counting chamber, dark lined