Sample vials, trident vials
Tall, 1,75 ml, 11×36 mm, Yes
212-7053
: MURRAY S
Vials made from neutral glass with PP screw cap. Designed for a wide variety of uses in the laboratory, production, more.
- Robust and economical, will withstand moderate temperature change
- Screw cap with foil coated insert for a better seal
- Easy handling
: For production related reasons, the number of vials per box may vary.
- Cap included:Yes
- Ø×H:11×36 mm
- Capacity:1,75 ml
- Description:Tall
- Seq:01



