Containers with screw cap
200 ml, Without label, Individually wrapped, PS
Catalog # ANICC200S120
Supplier: Anicrin
Jump to:
Product Details & Documents
Containers made of PS or PP with colour coded screw caps.
- Good chemical resistance
- Good temperature resistance
: Specify the cap colour (red, yellow and white) required.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Packed:Individually wrapped
- Version:Without label
- Capacity:200 ml
- Material:PS
- Seq:03