Jars with screw cap
with polypropylene screw cap, individually wrapped, labelled, 150 ml
Catalogus # 216-2051
Leverancier: Anicrin
Ga naar:
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
- Twist cap for quick opening and closing
- Very good temperature resistance
- Very good chemical resistance
Let op: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specificaties
- Type:with polypropylene screw cap, individually wrapped, labelled
- Capacity:150 ml
- Seq:07