Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
with polypropylene screw cap, irradiated, individually wrapped, 150 ml
Catalog # 216-2050
Supplier:   Anicrin
Jars with screw cap
Catalog # 216-2050
Supplier:  Anicrin
Jars with screw cap
Jars with screw cap
with polypropylene screw cap, irradiated, individually wrapped, 150 ml
Price
107,00 €
Log in for availability
1 * 250 ST
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Type:
    with polypropylene screw cap, irradiated, individually wrapped
  • Capacity:
    150 ml
  • Seq:
    06