ContactUsLinkComponent
to hold 96-well & 384-well plates w/locking rod, capacity = 84 plates w/lids or 96 plates w/out lids 22 L x 9 3/8 H x 5 1/2 W 1 * 1 items
Catalog # REVC7147
Supplier:  REVCO TECHNOLOGIES
to hold 96-well & 384-well plates w/locking rod, capacity = 84 plates w/lids or 96 plates w/out lids 22 L x 9 3/8 H x 5 1/2 W 1 * 1 items
Catalog # REVC7147
Supplier:  REVCO TECHNOLOGIES
Frequently Bought Together