Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ethidium bromide

Ethidium bromide

Glass bottle, 5 ml
Catalog # PIER17896
Toxic
Health
Ethidium bromide
Catalog # PIER17896
CAS Number:  1239-45-8
Ethidium bromide
Ethidium bromide

Glass bottle, 5 ml

CAS Number:  1239-45-8
Price
92,80 €
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0.1 mmHg)
  • Melting Pt:261…264 °C
  • Density:1.01 g/cm³ (20 °C)
  • Flash Pt:<100 °C
  • Storage Temperature:Ambient
  • EINECS:214-984-6
  • UN:2811
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.