Lid gasket CV Silicon 24 cm white single packed 1 * 1 items
Catalog # CERT1260404
Supplier: CERTOCLAV
Lid gasket CV Silicon 24 cm white single packed 1 * 1 items
Catalog # CERT1260404
Supplier: CERTOCLAV
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Media additives and supplements
Supplier: Merck
Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection