Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®NucleasEliminator™, Decontamination solution

VWR®NucleasEliminator™, Decontamination solution

NucleasEliminator™, ready to use liquid, 500 mL
Catalog # E891-500ML
Supplier:   VWR Chemicals
VWR®NucleasEliminator™, Decontamination solution
Catalog # E891-500ML
Supplier:  VWR Chemicals
CAS Number:  
VWR®NucleasEliminator™, Decontamination solution
VWR®NucleasEliminator™, Decontamination solution

NucleasEliminator™, ready to use liquid, 500 mL

CAS Number:  
Price
160,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces.

  • Effectively removes and deactivates nuclease contamination, even at high concentrations
  • Safe on all laboratory glass and plastic ware
  • Stable at room temperature for at least two years

Simply wipe, immerse, or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature, and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips, and electrophoresis units.

A 100 ml bottle of the ready to use liquid will treat up to 100 microcentrifuge tubes or 25 casting trays for agarose electrophoresis.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    NucleasEliminator™, ready to use liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.