Orders LinkContactUsLinkComponent
VWR®NucleasEliminator™, Decontamination solution
VWR®NucleasEliminator™, Decontamination solution
NucleasEliminator™, impregnated wipes, 25
Catalog # E891-25PK
Supplier:   VWR Chemicals
VWR®NucleasEliminator™, Decontamination solution
Catalog # E891-25PK
Supplier:  VWR Chemicals
CAS Number:  
VWR®NucleasEliminator™, Decontamination solution
VWR®NucleasEliminator™, Decontamination solution
NucleasEliminator™, impregnated wipes, 25
CAS Number:  
Price
102,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces.
  • Effectively removes and deactivates nuclease contamination, even at high concentrations
  • Safe on all laboratory glass and plastic ware
  • Stable at room temperature for at least two years
Simply wipe, immerse, or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature, and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips, and electrophoresis units.
A 100 ml bottle of the ready to use liquid will treat up to 100 microcentrifuge tubes or 25 casting trays for agarose electrophoresis.
Specifications
Specifications
  • Pk:
    25
  • Description:
    NucleasEliminator™, impregnated wipes