Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Buffer Standard Solutions

VWR® Buffer Standard Solutions

10,0, ±0,03, 25 °C, Plastic bottle, 500 mL
Catalog # E464-500ML
Supplier:   VWR Chemicals
VWR® Buffer Standard Solutions
Catalog # E464-500ML
Supplier:  VWR Chemicals
CAS Number:  
VWR® Buffer Standard Solutions
VWR® Buffer Standard Solutions

10,0, ±0,03, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
110,00 €
Log in for availability
1 * 500 mL
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,03
  • pH value:
    10,0

Specifications