Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Buffer Standard Solutions

VWR® Buffer Standard Solutions

4,00, ±0,03, 25 °C, Plastic bottle, 500 mL
Catalogus # E452-500ML
Leverancier:   VWR Chemicals
VWR® Buffer Standard Solutions
Catalogus # E452-500ML
Leverancier:  VWR Chemicals
CAS-nummer:  
VWR® Buffer Standard Solutions
VWR® Buffer Standard Solutions

4,00, ±0,03, 25 °C, Plastic bottle, 500 mL

CAS-nummer:  
Price
78,40 €
Log in voor beschikbaarheid
1 * 500 mL
Specificaties

Specificaties

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,03
  • pH value:
    4,00

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.