Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Sorbitol ≥96%, high purity

D-(-)-Sorbitol ≥96%, high purity

HDPE bottle, 500 g
Catalog # 0691-500G
Supplier:   VWR Chemicals
D-(-)-Sorbitol ≥96%, high purity
Catalog # 0691-500G
Supplier:  VWR Chemicals
CAS Number:  50-70-4
D-(-)-Sorbitol ≥96%, high purity
D-(-)-Sorbitol ≥96%, high purity

HDPE bottle, 500 g

CAS Number:  50-70-4
Price
244,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:98…100 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specification Test Results

  • Arsenic:
    <0.0003%
  • Chloride:
    <0.005%
  • Heavy Metals:
    <0.001%
  • Moisture (KF):
    ≤1%
  • Purity (Anhydrous):
    ≥96%
  • Residue after Ignition:
    <0.1%
  • Sulphate:
    <0.01%

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.