Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pasteur pipettes

2,0 ml, 7 mm, 225 mm, 250/box, 4 boxes/pack
  612-1719
 :   Brand
Pasteur pipettes
  612-1719
 :  Brand
Pasteur pipettes
Pasteur pipettes

2,0 ml, 7 mm, 225 mm, 250/box, 4 boxes/pack

 
88,60 €
   
1 * 1.000 ST
 

Soda-lime-glass.

Long drawn-out capillary tip. Suction end with constriction for a cotton plug. int.Ø of the tip: 1,0 mm.

 : ISO 7712

 
  • Ext.Ø:
    7 mm
  • Packed:
    250/box, 4 boxes/pack
  • Capacity:
    2,0 ml
  • Length:
    225 mm